missing translation for 'onlineSavingsMsg'
Learn More

Hexan-1-ol, Extra Pure, SLR, Fisher Chemical™

Product Code. 12628635 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 L
Packaging:
Metal can
Unit Size:
25 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12628635 25 L Metal can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12628635 Supplier Fisher Chemical Supplier No. H/0450/25

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 111-27-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number 2982
InChI Key ZSIAUFGUXNUGDI-UHFFFAOYSA-N
Synonym 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
PubChem CID 8103
ChEBI CHEBI:87393
IUPAC Name hexan-1-ol
SMILES CCCCCCO

Specifications

Melting Point -52°C
Color Colorless
Boiling Point 156°C
Packaging Metal can
Quantity 25 L
Formula Weight 102.18g/mol
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.