missing translation for 'onlineSavingsMsg'
Learn More

Hexaminolevulinate hydrochloride, MedChemExpress

Product Code. 16477046
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16477046

Brand: Medchem Express HY16045

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hexaminolevulinate hydrochloride is a fluorescent agent, has approved for cystoscopic detection of papillary bladder cancer.

Hexaminolevulinate hydrochloride is a fluorescent agent, has approved for cystoscopic detection of papillary bladder cancer.

Specifications

Chemical Name or Material Hexaminolevulinate hydrochloride
Physical Form Solid
CAS 140898-91-5
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C11H22ClNO3
For Use With (Application) Cancer-programmed cell death
Synonym Hexyl 5-aminolevulinate hydrochloride P-1206 5-Aminolevulinic acid hexyl ester hydrochloride
Solubility Information DMSO : ≥ 100 mg/mL (397.22 mM) ∣H2O : 100 mg/mL (397.22 mM; Need ultrasonic)
SMILES O=C(OCCCCCC)CCC(CN)=O.[H]Cl
Molecular Weight (g/mol) 251.75
Formula Weight 251.75
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.