missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamidine Impurity A Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Hexamidine Impurity A Hydrochloride |
| Analyte or Component Names | Hexamidine Impurity A Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H25 N3 O3 . H Cl |
| Synonym | 4-((6-(4-Carbamimidoylphenoxy)hexyl)oxy)benzamide Hydrochloride |
| InChI Formula | InChI=1S/C20H25N3O3.ClH/c21-19(22)15-5-9-17(10-6-15)25-13-3-1-2-4-14-26-18-11-7-16(8-12-18)20(23)24;/h5-12H,1-4,13-14H2,(H3,21,22)(H2,23,24);1H |
| Solution Type | Neat |
| SMILES | Cl.NC(=N)c1ccc(OCCCCCCOc2ccc(cc2)C(=O)N)cc1 |
| IUPAC Name | 4-[6-(4-carbamimidoylphenoxy)hexoxy]benzamide;hydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?