missing translation for 'onlineSavingsMsg'
Learn More

Hexamidine-d12 Dihydrochloride, TRC

Product Code. 30396309
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396309 1 mg 1mg
30398207 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30396309 Supplier Toronto Research Chemicals Supplier No. TRCH2942071MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1286461-12-8
Molecular Formula C20 D12 H14 N4 O2
Molecular Weight (g/mol) 366.52
Synonym 4,4'-[1,6-(Hexane-d12)diylbis(oxy)]bisbenzenecarboximidamide Hydrochloride, 1,6-Bis(4-amidinophenoxy)hexane-d12 Hydrochloride, 4,4-Diamidino-1,6-diphenoxyhexane-d12 Hydrochloride, 4,4'-(Hexamethylenedioxy)dibenzamidine-d12 Hydrochloride, Desmodine-d12 Hydrochloride
IUPAC Name 4-[6-(4-carbamimidoylphenoxy)-1,1,2,2,3,3,4,4,5,5,6,6-dodecadeuteriohexoxy]benzenecarboximidamide
SMILES [2H]C([2H])(Oc1ccc(cc1)C(=N)N)C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])Oc2ccc(cc2)C(=N)N

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C20H26N4O2/c21-19(22)15-5-9-17(10-6-15)25-13-3-1-2-4-14-26-18-11-7-16(8-12-18)20(23)24/h5-12H,1-4,13-14H2,(H3,21,22)(H3,23,24)/i1D2,2D2,3D2,4D2,13D2,14D2
Formula Weight 366.281
Chemical Name or Material Hexamidine-d12 Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.