missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylenimine, TRC

Product Code. 30395340
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30395340 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395340 Supplier Toronto Research Chemicals Supplier No. TRCH29423510G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexamethylenimine
CAS 111-49-9
Molecular Formula C6H13N
Quantity 10 g
Synonym 1-Azacycloheptane, Azacycloheptane, Azepan, Azepane, G 0, Hexahydro-1H-azepine, Hexahydroazepine, Homopiperidine, NSC 16236, Perhydroazepine
InChI Formula InChI=1S/C6H13N/c1-2-4-6-7-5-3-1/h7H,1-6H2
SMILES C1CCCNCC1
IUPAC Name azepane
Molecular Weight (g/mol) 99.17
Formula Weight 99.1048
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.