missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylenetetramine, TRC

Product Code. 17967929
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17967929 10 g 10g
17958839 50 mg 50mg
17903945 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 17967929 Supplier Toronto Research Chemicals Supplier No. TRCH29422010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 100-97-0
Molecular Formula C6 H12 N4
Molecular Weight (g/mol) 140.19
Synonym Methenamine, 1,3,5,7-Tetraazotricyclo[3.3.1.13,7]decane, Hexamethylenetetramine (8CI), 1,3,5,7-Tetraazaadamantane, 1,3,5,7-Tetraazatricyclo[3.3.1.13,7]decane (Urotropine), Aceto HMT, Aminoform, Ammoform, Antihydral, Cohedur H 30, Cystogen, Duirexol, EL 10, EL 10 (corrosion inhibitor), Ekagom H, Formamine, Formin, Formin (heterocycle), HA, HMT, HMTA, Heksa K, Herax UTS, Heterin, Hexa, Hexa (vulcanization accelerator), Hexa B, Hexa-Flo-Pulver, Hexaform, Hexamethylenamine, Hexamine, Hexamine (heterocycle), Hexamine Superfine, Hexasan, Methenamide, Methenamin, Methenamine, NSC 26346, NSC 403347, Nocceler H, Nocceler H-PO, Preparation AF, Rhenogran HEXA 80, S 4, S 4 (heterocycle), Sanceler H, Sanceler HT, Sanceler HT-PO, Thixon 715B, Uramin, Uratrine, Uritone, Urodeine, Urotropin, Urotropine, Vulkacit H, Vulkacit H 30, Xametrin
IUPAC Name 1,3,5,7-Tetraazotricyclo[3.3.1.13,7]decane
SMILES C1N2CN3CN(CN1C3)C2

Especificaciones

Quantity 10 g
InChI Formula InChI=1S/C6H12N4/c1-7-2-9-4-8(1)5-10(3-7)6-9/h1-6H2
Formula Weight 140.1062
Chemical Name or Material Urotropine
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.