missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylenetetramine, ≥99%, Reagent grade, For ACS analysis, MP Biomedicals™

Product Code. 11430622 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430622 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11430622 Supplier MP Biomedicals Supplier No. 0219405091
  • ONLINE EXCLUSIVE PRICE
    £146.00
    £136.50 / 1kg
    Save £9.50
    6% Off
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Specifications

CAS 100-97-0
Color White
Density 1.33g/cm3 at 20°C
Molecular Formula C6H12N4
Quantity 1 kg
Synonym methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
InChI Key VKYKSIONXSXAKP-UHFFFAOYSA-N
SMILES C1N2CN3CN1CN(C2)C3
Molecular Weight (g/mol) 140.19
PubChem CID 4101
ChEBI CHEBI:6824
Formula Weight 140.2
Percent Purity ≥99%
Grade Reagent
Physical Form Crystalline Powder
Chemical Name or Material Hexamethylenetetramine, For ACS analysis
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hexamethylenetetramine
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H228-Flammable solid.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: hexamethylenetetramine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.