missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamethylenediamine-N,N,N',N'-tetrakis(methylphosphonic Acid), TRC
Specifications
Specifications
| Chemical Name or Material | Hexamethylenediamine-N,N,N',N'-tetrakis(methylphosphonic Acid) |
| CAS | 23605-74-5 |
| Molecular Formula | C10 H28 N2 O12 P4 |
| Quantity | 1 g |
| Synonym | Hexanediylbis[nitrilobis(methylene)]]tetrakis[phosphonic Acid], (Hexamethylenedinitrilo)tetrakis[methylenephosphonic Acid], 1,6-Hexanediaminetetrakis(methylene phosphonic Acid), Briquest 462, Dequest 2051, HDTMP, HDTMPA, Hexamethylenedinitrilotetrakis(methylphosphonic Acid), Hexanediaminetetra(methylenephosphonic Acid), N,N,N',N'-1,6-Hexamethylenediaminetetrakis(methylenephosphonic Acid), N,N,N',N'-Hexamethylenediaminetetra(methylenephosphonic Acid) |
| InChI Formula | InChI=1S/C10H28N2O12P4/c13-25(14,15)7-11(8-26(16,17)18)5-3-1-2-4-6-12(9-27(19,20)21)10-28(22,23)24/h1-10H2,(H2,13,14,15)(H2,16,17,18)(H2,19,20,21)(H2,22,23,24) |
| SMILES | OP(=O)(O)CN(CCCCCCN(CP(=O)(O)O)CP(=O)(O)O)CP(=O)(O)O |
| IUPAC Name | [6-[bis(phosphonomethyl)amino]hexyl-(phosphonomethyl)amino]methylphosphonic acid |
| Molecular Weight (g/mol) | 492.231 |
| Formula Weight | 492.059 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?