missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylenediamine-N,N,N',N'-tetrakis(methylphosphonic Acid), TRC

Product Code. 30398547
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398547 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398547 Supplier Toronto Research Chemicals Supplier No. TRCH294326500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexamethylenediamine-N,N,N',N'-tetrakis(methylphosphonic Acid)
CAS 23605-74-5
Molecular Formula C10 H28 N2 O12 P4
Quantity 500 mg
Synonym Hexanediylbis[nitrilobis(methylene)]]tetrakis[phosphonic Acid], (Hexamethylenedinitrilo)tetrakis[methylenephosphonic Acid], 1,6-Hexanediaminetetrakis(methylene phosphonic Acid), Briquest 462, Dequest 2051, HDTMP, HDTMPA, Hexamethylenedinitrilotetrakis(methylphosphonic Acid), Hexanediaminetetra(methylenephosphonic Acid), N,N,N',N'-1,6-Hexamethylenediaminetetrakis(methylenephosphonic Acid), N,N,N',N'-Hexamethylenediaminetetra(methylenephosphonic Acid)
InChI Formula InChI=1S/C10H28N2O12P4/c13-25(14,15)7-11(8-26(16,17)18)5-3-1-2-4-6-12(9-27(19,20)21)10-28(22,23)24/h1-10H2,(H2,13,14,15)(H2,16,17,18)(H2,19,20,21)(H2,22,23,24)
SMILES OP(=O)(O)CN(CCCCCCN(CP(=O)(O)O)CP(=O)(O)O)CP(=O)(O)O
IUPAC Name [6-[bis(phosphonomethyl)amino]hexyl-(phosphonomethyl)amino]methylphosphonic acid
Molecular Weight (g/mol) 492.231
Formula Weight 492.059
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.