missing translation for 'onlineSavingsMsg'
Learn More

Hexamethyldisiloxane, TRC

Product Code. 30134773
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134773 10 g 10g
17921565 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134773 Supplier Toronto Research Chemicals Supplier No. TRCH29421510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-46-0
Molecular Formula C6H18OSi2
Molecular Weight (g/mol) 162.38
Synonym HMDO, 1,1,1,3,3,3-Hexamethyldisiloxane, Belsil DM 0.65, Bis(trimethylsilyl) ether, Bis(trimethylsilyl) oxide, DC 0.65cs200, Dow Corning 200/0.65, Dow Corning OS 10, Ethoxytrimethylsilane-hexamethyldisiloxane copolymer, Glipoxan, HMDS, HMDSO, HX 18, Hexamethyldisiloxane, KF 96L0.65, LS 7130, MM, MM (siloxane), NSC 43346, OL-AK 065, OS 10, OS 10 (siloxane), PSF-0.65cSt, SF 0.65, SH 200-0.65CS, SH 200-0.65cSt, SH 200-0.6CS, SWS-F 221, Volasil DM 0.65, Volasil DM 0.65SP
IUPAC Name trimethyl(trimethylsilyloxy)silane
SMILES C[Si](C)(C)O[Si](C)(C)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H18OSi2/c1-8(2,3)7-9(4,5)6/h1-6H3
Formula Weight 162.09
Chemical Name or Material Hexamethyldisiloxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.