missing translation for 'onlineSavingsMsg'
Learn More

Hexamethyldisiloxane, 99.3%, MP Biomedicals™

Product Code. 11463801 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463801 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11463801 Supplier MP Biomedicals Supplier No. 0215124980
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 107-46-0
Color Colorless
Density 0.76g/mL
Molecular Formula C6H18OSi2
Quantity 100 g
Synonym hexamethyldisiloxane, disiloxane, hexamethyl, hexamethyl disiloxane, oxybis trimethylsilane, fluka ag, hmdso, bis trimethylsilyl ether, belsil dm 0.65, bis trimethylsilyl oxide
InChI Key UQEAIHBTYFGYIE-UHFFFAOYSA-N
SMILES C[Si](C)(C)O[Si](C)(C)C
IUPAC Name trimethyl(trimethylsilyloxy)silane
Molecular Weight (g/mol) 162.379
PubChem CID 24764
ChEBI CHEBI:78002
Formula Weight 162.4
Percent Purity 99.3%
Physical Form Liquid
Chemical Name or Material Hexamethyldisiloxane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.