missing translation for 'onlineSavingsMsg'
Learn More

Hexamethyldisilazane, TRC

Product Code. 30395233
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30395233 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395233 Supplier Toronto Research Chemicals Supplier No. TRCH296700500G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexamethyldisilazane
CAS 999-97-3
Purity Grade Notes GC
Molecular Formula C6H19NSi2
Quantity 500 g
Synonym 1,1,1,3,3,3-Hexamethyldisilazane, 1,1,1-Trimethyl-N-(trimethylsilyl)silanamine, A 166, A 166 (Silazane), Bis(trimethylsilyl)amine, Di(trimethylsilyl)amine, Dynasylan HMDS, HMD 3, HMDS, HMDS (silazane), HMDS 3, Hexamethyldisilazane, Hexamethyldisilylamine, LS 7150, NSC 93895, OAP, SE 31, SE 31 (silazane), SIH 6110.1, SZ 6079, TSL 8802, TSR 8802, Z 6079
InChI Formula InChI=1S/C6H19NSi2/c1-8(2,3)7-9(4,5)6/h7H,1-6H3
SMILES C[Si](C)(C)N[Si](C)(C)C
IUPAC Name [dimethyl-(trimethylsilylamino)silyl]methane
Molecular Weight (g/mol) 161.39
Formula Weight 161.1056
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.