missing translation for 'onlineSavingsMsg'
Learn More

Hexamethonium Bromide, TRC

Product Code. 30403923
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 30403923

Brand: Toronto Research Chemicals TRCH29670825G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexamethonium Bromide
CAS 55-97-0
Molecular Formula C12H30Br2N2
Quantity 25 g
Synonym 1,6-Hexanebis(trimethylammonium) Dibromide, 1,6-Bis(trimethylammonium)hexane Dibromide, Bistrium Bromide, Esametina, Gangliostat, Hexamethonium bromide, Methobromin, Methobromine, N,N,N,N',N',N'-Hexamethyl(hexamethylene)diammonium bromide, Simpatoblock, Vegolysen, Vegolysin
InChI Formula InChI=1S/C12H30N2.2BrH/c1-13(2,3)11-9-7-8-10-12-14(4,5)6;;/h7-12H2,1-6H3;2*1H/q+2;;/p-2
SMILES [Br-].[Br-].C[N+](C)(C)CCCCCC[N+](C)(C)C
IUPAC Name trimethyl-[6-(trimethylazaniumyl)hexyl]azanium;dibromide
Molecular Weight (g/mol) 362.19
Formula Weight 360.0776
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.