missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamethonium Bromide, MedChemExpress
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
Hexamethonium Bromide is a non-selective ganglionic nicotinic-receptor antagonist (nAChR) antagonist, with mixed competitive and noncompetitive activity. Hexamethonium Bromide has anti-hypertensive activity. Hexamethonium Bromide attenuates sympathetic activity and blood pressure in spontaneously hypertensive animal models.
Specifications
Specifications
| Chemical Name or Material | Hexamethonium Bromide |
| Physical Form | Solid |
| CAS | 55-97-0 |
| Color | White |
| Quantity | 1 g |
| Purity Grade Notes | Research |
| Molecular Formula | C12H30Br2N2 |
| For Use With (Application) | Neuroscience-Neurodegeneration |
| Solubility Information | H2O : ≥ 100 mg/mL (276.10 mM) ∣DMSO : 16.67 mg/mL (46.03 mM; Need ultrasonic) |
| SMILES | C[N+](C)(C)CCCCCC[N+](C)(C)C.[Br-].[Br-] |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction