missing translation for 'onlineSavingsMsg'
Learn More

Hexamethonium bromide, 98+%

Product Code. 15433239
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15433239

Brand: Thermo Scientific Alfa Aesar J60466.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nicotinic acetyl choline receptor antagonist | CAS: 55-97-0 | C12H30Br2N2 | 362.194 g/mol

Nicotinic acetyl choline receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-97-0
Molecular Formula C12H30Br2N2
Molecular Weight (g/mol) 362.194
MDL Number MFCD00011787
InChI Key FAPSXSAPXXJTOU-UHFFFAOYSA-L
Synonym hexamethonium bromide, hexamethonium dibromide, simpatoblock, gangliostat, esametina, hexameton, vegolysen, vegolysin, bistrium bromide, hexonium dibromide
PubChem CID 5938
IUPAC Name trimethyl-[6-(trimethylazaniumyl)hexyl]azanium;dibromide
SMILES C[N+](C)(C)CCCCCC[N+](C)(C)C.[Br-].[Br-]

Specifications

Melting Point 282°C to 285°C (decomposition)
Color White
Linear Formula (CH3)3N(Br)(CH2)6N(Br)(CH3)3
Quantity 25 g
Beilstein 3715749
Merck Index 14,4688
Formula Weight 362.19
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Hexamethonium bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.