missing translation for 'onlineSavingsMsg'
Learn More

Hexametazidine, TRC

Product Code. 30135579
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135579 2.5 g 2.5g
30005862 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135579 Supplier Toronto Research Chemicals Supplier No. TRCH2911502.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1257-19-8
Molecular Formula C24 H34 N2 O6
Molecular Weight (g/mol) 446.54
Synonym 1,4-Bis(2,3,4-trimethoxybenzyl)piperazine, Trimetazidine Dihydrochloride Imp. B (EP)
IUPAC Name 1,4-bis[(2,3,4-trimethoxyphenyl)methyl]piperazine
SMILES COc1ccc(CN2CCN(Cc3ccc(OC)c(OC)c3OC)CC2)c(OC)c1OC

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C24H34N2O6/c1-27-19-9-7-17(21(29-3)23(19)31-5)15-25-11-13-26(14-12-25)16-18-8-10-20(28-2)24(32-6)22(18)30-4/h7-10H,11-16H2,1-6H3
Formula Weight 446.2417
Chemical Name or Material 1,4-Bis(2,3,4-trimethoxybenzyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.