missing translation for 'onlineSavingsMsg'
Learn More

Hexaketocyclohexane octahydrate, 99%

Product Code. 11327456
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327456 10 g 10g
11317456 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327456 Supplier Thermo Scientific Alfa Aesar Supplier No. L09488.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7255-28-9 | C6O6 | 168.06 g/mol

Hexaketocyclohexane octahydrate is used in the preparation of hexaazatriphenylenehexacarbonitrile. It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexaketocyclohexane octahydrate is used in the preparation of hexaazatriphenylenehexacarbonitrile. It is used as pharmaceutical intermediates.

Notes
Store at 4°C. Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7255-28-9
Molecular Formula C6O6
Molecular Weight (g/mol) 168.06
MDL Number MFCD00001652,MFCD00149074
InChI Key PKRGYJHUXHCUCN-UHFFFAOYSA-N
Synonym cyclohexanehexone, triquinoyl, cyclohexane-1,2,3,4,5,6-hexaone, cyclohexanehexaone, hexaketocyclohexane, unii-7zr8062lfd, trichinoyl, hexaoxocyclohexane, hexaoxocyclohexaneoctahydrate, cyclohexane-hexone, octahydrate
PubChem CID 68240
IUPAC Name cyclohexane-1,2,3,4,5,6-hexone
SMILES O=C1C(=O)C(=O)C(=O)C(=O)C1=O

Specifications

Melting Point 90°C to 93°C
Quantity 10 g
Beilstein 2091040
Formula Weight 312.20 (168.07 Anhydrous)
Percent Purity 99%
Chemical Name or Material Hexaketocyclohexane octahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.