missing translation for 'onlineSavingsMsg'
Learn More

Hexaketocyclohexane octahydrate, 97%, Thermo Scientific Chemicals

Product Code. 10031730 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10031730 10 g Glass bottle 10g
10764281 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10031730 Supplier Thermo Scientific Acros Supplier No. 156280100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7255-28-9
Molecular Formula C6O6
Molecular Weight (g/mol) 168.06
MDL Number MFCD00001652,MFCD00149074
InChI Key PKRGYJHUXHCUCN-UHFFFAOYSA-N
Synonym cyclohexanehexone, triquinoyl, cyclohexane-1,2,3,4,5,6-hexaone, cyclohexanehexaone, hexaketocyclohexane, unii-7zr8062lfd, trichinoyl, hexaoxocyclohexane, hexaoxocyclohexaneoctahydrate, cyclohexane-hexone, octahydrate
PubChem CID 68240
IUPAC Name cyclohexane-1,2,3,4,5,6-hexone
SMILES O=C1C(=O)C(=O)C(=O)C(=O)C1=O

Specifications

Melting Point 90.0°C to 93.0°C
Color Beige to Brown-Gray
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 10 g
Beilstein 07, 907
Formula Weight 312.17
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Hexaketocyclohexane octahydrate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Hexaketocyclohexane octahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.