missing translation for 'onlineSavingsMsg'
Learn More

Hexahydrocurcumin, TRC

Product Code. 30138236
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138236 500 mg 500mg
30005320 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138236 Supplier Toronto Research Chemicals Supplier No. TRCH281370500MG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36062-05-2
Molecular Formula C21 H26 O6
Molecular Weight (g/mol) 374.43
Synonym 3-Heptanone, 5-hydroxy-1,7-bis(4-hydroxy-3-methoxyphenyl)-, 5-Hydroxy-1,7-bis(4-hydroxy-3-methoxyphenyl)-3-heptanone, Hexahydrocurcumin
IUPAC Name 5-hydroxy-1,7-bis(4-hydroxy-3-methoxyphenyl)heptan-3-one
SMILES COc1cc(CCC(O)CC(=O)CCc2ccc(O)c(OC)c2)ccc1O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C21H26O6/c1-26-20-11-14(5-9-18(20)24)3-7-16(22)13-17(23)8-4-15-6-10-19(25)21(12-15)27-2/h5-6,9-12,16,22,24-25H,3-4,7-8,13H2,1-2H3
Formula Weight 374.1729
Chemical Name or Material Hexahydrocurcumin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.