missing translation for 'onlineSavingsMsg'
Learn More

Hexahydrocurcumin-d6, TRC

Product Code. 30391153
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391153 5 mg 5mg
30399847 10 mg -
30404213 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30391153 Supplier Toronto Research Chemicals Supplier No. TRCH2812775MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C21H20D6O6
Molecular Weight (g/mol) 380.46
Synonym 5-Hydroxy-1,7-bis(4-hydroxy-3-methoxyphenyl)-3-heptanone-d6
SMILES OC1=C(OC([2H])([2H])[2H])C=C(CCC(CC(O)CCC2=CC=C(O)C(OC([2H])([2H])[2H])=C2)=O)C=C1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H26O6/c1-26-20-11-14(5-9-18(20)24)3-7-16(22)13-17(23)8-4-15-6-10-19(25)21(12-15)27-2/h5-6,9-12,16,22,24-25H,3-4,7-8,13H2,1-2H3/i1D3,2D3
Chemical Name or Material Hexahydrocurcumin-d6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.