missing translation for 'onlineSavingsMsg'
Learn More

Hexahydro-4-methylphthalic Anhydride, TRC

Product Code. 30402388
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
5g
Product Code. Quantity unitSize
30402388 25 g -
30397021 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 30402388

Brand: Toronto Research Chemicals TRCH29378525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19438-60-9
Molecular Formula C9 H12 O3
Molecular Weight (g/mol) 168.19
Synonym 1,3-Isobenzofurandione, hexahydro-5-methyl-, 1,2-Cyclohexanedicarboxylic anhydride, 4-methyl- (8CI), Hexahydro-5-methyl-1,3-isobenzofurandione, 1-Methylcyclohexane-4,5-Dicarboxylic anhydride, 4-Methyl-1,2-cyclohexanedicarboxylic anhydride, 4-Methylhexahydrophthalic acid anhydride, 4-Methylhexahydrophthalic anhydride, HN 7000, Hexahydro-4-methylphthalic anhydride, Lindride 52, NSC 128883, Quinhard HH 400, Rikacid MH, Rikacid MH-T, WH 700
IUPAC Name 5-methyl-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione
SMILES CC1CCC2C(C1)C(=O)OC2=O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H12O3/c1-5-2-3-6-7(4-5)9(11)12-8(6)10/h5-7H,2-4H2,1H3
Formula Weight 168.0786
Chemical Name or Material 4-Methyl-1,2-cyclohexanedicarboxylic acid anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.