missing translation for 'onlineSavingsMsg'
Learn More

Hexahydro-1,3-isobenzofurandione-d8, TRC

Product Code. 30400719
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400719 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30400719 Supplier Toronto Research Chemicals Supplier No. TRCH294022250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexahydro-1,3-isobenzofurandione-d8
CAS 89614-24-4
Molecular Formula C8H2D8O3
Quantity 250 mg
Synonym 1,2-Cyclohexanedicarboxylic Anhydride-d8, 1,2-Cyclohexanedicarboxylic Acid Anhydride-d8, Aradur HY 925-d8, Araldite HT 904-d8, Araldite HT 907-d8, Araldite HY 907-d8, Araldite HY 925-d8, Araldite Hardener HY 925-d8, Cyclohexane-1,2-dicarboxylic Acid Anhydride-d8, Cyclohexanedicarboxylic Anhydride-d8, EP 400B-d8, Epilox H 11-01-d8, HHPA-d8, HT 907-d8, HY 925-d8, Hexahydro-1,3-isobenzofurandione-d8, Hexahydro-2-benzofuran-1,3-dione-d8, Hexahydrophthalic Acid Anhydride-d8, Hexahydrophthalic Anhydride-d8, Lekutherm Hardener H-d8, MH 700E-d8, NSC 8622-d8, Rikacid HH-d8, Rikacid HH-A-d8, Rikacid HHPA-d8, Rikacid MH 700E-d8, Rutadur AG-d8, Rutapox HX-d8
InChI Formula InChI=1S/C8H10O3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h5-6H,1-4H2/i1D2,2D2,3D2,4D2
SMILES O=C1OC(C2C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C21)=O
IUPAC Name hexahydroisobenzofuran-1,3-dione-4,4,5,5,6,6,7,7-d8
Molecular Weight (g/mol) 162.21
Formula Weight 162.11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.