missing translation for 'onlineSavingsMsg'
Learn More

Hexafluorosilicic acid, 35% w/w aq. soln.

Product Code. 11383696
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Conditionnement:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity unitSize
11383696 500 g 500g
11373696 100 g 100g
11393696 2500 g 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11383696 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur L14829.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 16961-83-4 | F6H2Si | 144.091 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Hexafluorosilicic acid is commonly used as a source of fluoride. It is converted to a variety of useful hexafluorosilicate salts. It is also used as an electrolyte in the Betts electrolytic process for refining lead. It is an important organic reagent for cleaving Si-O bonds of silyl ethers. Further, it is used as wood a preservation agent and also used in surface modification of calcium carbonate.

Solubility
Miscible with water.

Notes
Incompatible in strong oxidizing agents, metals, alkalis, strong acids, stoneware and glass.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 16961-83-4
Molecular Formula F6H2Si
Molecular Weight (g/mol) 144.091
MDL Number MFCD00036289
InChI Key OHORFAFFMDIQRR-UHFFFAOYSA-P
Synonym hexafluorosilicic acid, fluorosilicic acid, dihydrogen hexafluorosilicate, silicate 2-, hexafluoro-, dihydrogen, hexafluorosilicon 2-; hydron, dihydrogen hexafluorosilanediuide, silicofluoridd, hexafluorosilic acid, hydrofluorosilic acid, kieselfluorwasserstoffsaure
PubChem CID 21863527
IUPAC Name hexafluorosilicon(2-);hydron
SMILES [H+].[H+].F[Si-2](F)(F)(F)(F)F

Spécification

Boiling Point 108°C to 109°C
Quantity 500 g
Linear Formula H2SiF6
UN Number UN1778
Merck Index 14,4182
Solubility Information Miscible with water.
Formula Weight 144.09
Concentration or Composition (by Analyte or Components) 35% w/w aq. soln.
Density 1.32
Chemical Name or Material Hexafluorosilicic acid
compliance-icons
Identifiant produit
  • Hexafluorosilicic acid, 35% w/w aqueous solution
Mention d'avertissement
  • Danger
Catégorie de danger
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Mention de danger
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Conseils de prudence
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Informations supplémentaires
  • MIXTURE LIST-Contain: Fluorosilicic acid

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.