missing translation for 'onlineSavingsMsg'
Learn More

Hexafluorosilicic acid, 23% w/w aq. soln., Thermo Scientific Chemicals

Product Code. 11376319
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11376319 2.5 kg 2.5kg
11366319 100 g 100g
11354799 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11376319 Supplier Thermo Scientific Alfa Aesar Supplier No. 069109.A4
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
This item is not returnable. View return policy

It is mainly produced as a precursor to aluminum trifluoride and synthetic cryolite. It is commonly used as a source of fluoride for water fluoridation. Hexafluorosilicic acid is also converted to a variety of useful hexafluorosilicate salts. The potassium salt is used in the production of porcelains, the magnesium salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16961-83-4
Molecular Formula F6H2Si
Molecular Weight (g/mol) 144.091
MDL Number MFCD00036289
InChI Key OHORFAFFMDIQRR-UHFFFAOYSA-P
Synonym hexafluorosilicic acid, fluorosilicic acid, dihydrogen hexafluorosilicate, silicate 2-, hexafluoro-, dihydrogen, hexafluorosilicon 2-; hydron, dihydrogen hexafluorosilanediuide, silicofluoridd, hexafluorosilic acid, hydrofluorosilic acid, kieselfluorwasserstoffsaure
PubChem CID 21863527
IUPAC Name hexafluorosilicon(2-);hydron
SMILES [H+].[H+].F[Si-2](F)(F)(F)(F)F

Specifications

Color White through Yellow
Physical Form Liquid
Quantity 2.5 kg
UN Number UN1778
Merck Index 14,4182
Formula Weight 144.09
Concentration or Composition (by Analyte or Components) 23% w/w aq. soln.
Density 1.223g/mL
Chemical Name or Material Hexafluorosilicic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.