missing translation for 'onlineSavingsMsg'
Learn More

Hexafluorobenzene, 99%

Product Code. 11460413
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460413 10 g 10g
11470413 50 g 50g
11480413 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460413 Supplier Thermo Scientific Alfa Aesar Supplier No. A11500.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 392-56-3 | C6F6 | 186.056 g/mol

Hexafluorobenzene is used as a solvent in photochemical reactions. It is also used as a reference compound in fluorine-19 NMR, carbon-13 NMR. It is used as a solvent in proton NMR, IR spectrum and UV-spectra. It is used as anticorrosive, antifriction and anti-tumor agents. Further, it is used as a reference molecule to investigate tissue oxygenation studies. It forms series of 1:1 complexes with naphthalene, anthracene, phenanthrene, pyrene and triphenylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexafluorobenzene is used as a solvent in photochemical reactions. It is also used as a reference compound in fluorine-19 NMR, carbon-13 NMR. It is used as a solvent in proton NMR, IR spectrum and UV-spectra. It is used as anticorrosive, antifriction and anti-tumor agents. It forms series of 1:1 complexes with naphthalene, anthracene, phenanthrene, pyrene and triphenylene.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 392-56-3
Molecular Formula C6F6
Molecular Weight (g/mol) 186.056
MDL Number MFCD00000288
InChI Key ZQBFAOFFOQMSGJ-UHFFFAOYSA-N
Synonym hexafluorobenzene, perfluorobenzene, benzene, hexafluoro, hexafluorbenzol, unii-cmc18t611k, benzene, 1,2,3,4,5,6-hexafluoro, hexa fluorobenzene, hexafluoro benzene, pubchem18879, acmc-1bmus
PubChem CID 9805
ChEBI CHEBI:38589
IUPAC Name 1,2,3,4,5,6-hexafluorobenzene
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)F

Specifications

Melting Point 4°C to 5°C
Density 1.616
Boiling Point 80°C to 81°C
Flash Point 10°C (50°F)
Assay Percent Range 99%
Refractive Index 1.378
Quantity 10 g
Beilstein 1683438
Solubility Information Immiscible with water.
Formula Weight 186.06
Physical Form Liquid
Chemical Name or Material Hexafluorobenzene
Show More Show Less
compliance-icons
Product Identifier
  • Hexafluorobenzene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Supplemental information
  • MIXTURE LIST-Contain: Hexafluorobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.