missing translation for 'onlineSavingsMsg'
Learn More

Hexafluoroacetone trihydrate, 98%

Product Code. 11392986
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392986 25 g 25g
11382986 5 g 5g
11302996 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11392986 Supplier Thermo Scientific Alfa Aesar Supplier No. L10777.14
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 34202-69-2 | C3H6F6O4 | 220.067 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34202-69-2
Molecular Formula C3H6F6O4
Molecular Weight (g/mol) 220.067
MDL Number MFCD00149061
InChI Key SNZAEUWCEHDROX-UHFFFAOYSA-N
Synonym hexafluoroacetone trihydrate, 1,1,1,3,3,3-hexafluoropropan-2-one trihydrate, 2-propanone, hexafluoro-, trihydrate, hfa trihydrate, acetone, hexafluoro-, trihydrate, 2-propanone, 1,1,1,3,3,3-hexafluoro-, trihydrate, perfluoroacetone trihydrate, hexafluoro-2-propanone trihydrate, hexafluoro-2-propanone trihydrate; perfluoroacetone trihydrate, hexafluoroacetonetrihydrate
PubChem CID 36719
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-one;trihydrate
SMILES C(=O)(C(F)(F)F)C(F)(F)F.O.O.O

Specifications

Melting Point 18°C to 21°C
Density 1.579
Boiling Point 105°C to 106°C
Refractive Index 1.319
Quantity 25 g
UN Number UN2552
Beilstein 607236
Formula Weight 220.07 (166.02 Anhydrous)
Percent Purity 98%
Chemical Name or Material Hexafluoroacetone trihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.