missing translation for 'onlineSavingsMsg'
Learn More

Hexaethyl phosphorous triamide, 97%

Product Code. 11494114
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494114 5 g 5g
11404124 25 g 25g
11414124 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494114 Supplier Thermo Scientific Alfa Aesar Supplier No. A12490.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2283-11-6 | C12H30N3P | 247.37 g/mol

Hexaethyl phosphorous triamide is used in the Barton syntheses of hindered alkenes, involving desulfurization of small-ring heterocycles with elimination of nitrogen or carbon dioxide. It is also used as a reagent for selective vicinal dehalogenation of halogenated alkanes to give perhalogenated alkenes. The method has potential for the synthesis of highly fluorinated and mixed fluoro-halo alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexaethyl phosphorous triamide is used in the Barton syntheses of hindered alkenes, involving desulfurization of small-ring heterocycles with elimination of nitrogen or carbon dioxide. It is also used as a reagent for selective vicinal dehalogenation of halogenated alkanes to give perhalogenated alkenes. The method has potential for the synthesis of highly fluorinated and mixed fluoro-halo alkenes.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2283-11-6
MDL Number MFCD00009041

Specifications

Density 0.903
Boiling Point 245°C to 246°C
Flash Point 58°C (136°F)
Molecular Formula C8H4F3NO
Refractive Index 1.4737
Quantity 5 g
UN Number UN1993
Beilstein 636187
Sensitivity Air Sensitive
Synonym 2-trifluoromethyl phenyl isocyanate, 1-isocyanato-2-trifluoromethyl benzene, benzene, 1-isocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isocyanate, o-trifluoromethylphenylisocyanate, isocyanic acid, alpha,alpha,alpha-trifluoro-o-tolyl ester, o-trifluoromethyl phenyl isocyanate, 2-trifluoromethyl phenylisocyanate, 2-trifluoromethyl benzenisocyanate, isocyanic acid, .alpha.,.alpha.,.alpha.-trifluoro-o-tolyl ester
Solubility Information Insoluble in water.
InChI Key GZWGTVZRRFPVAS-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)C(F)(F)F)N=C=O
IUPAC Name 1-isocyanato-2-(trifluoromethyl)benzene
Molecular Weight (g/mol) 187.12
PubChem CID 16794
Formula Weight 247.37
Percent Purity 97%
Chemical Name or Material Hexaethyl phosphorous triamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.