missing translation for 'onlineSavingsMsg'
Learn More

Hexadimethrine bromide, MedChemExpress

Product Code. 16430987
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16430987 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16430987 Supplier Medchem Express Supplier No. HY112735
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Hexadimethrine bromide (Polybrene) is a cationic polymer used routinely to enhance the efficiency of retrovirus vector-mediated gene transfer.

Hexadimethrine bromide (Polybrene) is a cationic polymer used routinely to enhance the efficiency of retrovirus vector-mediated gene transfer.

Specifications

Chemical Name or Material Hexadimethrine bromide
Color White
CAS 28728-55-4
Physical Form Solid
Purity Grade Notes Research
Molecular Formula (C13H30N2)n.Br2
Synonym Polybrene 1,5-Dimethyl-1,5-diazaundecamethylene polymethobromide
Solubility Information H2O : ≥ 125 mg/mL ∣DMSO : 5 mg/mL (Need ultrasonic and warming)
Quantity 50 mg
SMILES C[N+](CCCCCCC)(C)CCC[N+](C)(C)C.[Br-].[Br-].[n]
For Use With (Application) Cancer-programmed cell death
Grade Research
Show More Show Less
compliance-icons
Product Identifier
  • Hexadimethrine bromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Polybrene; 1,5-Dimethyl-1,5-diazaundecamethylene polymethobromide

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.