missing translation for 'onlineSavingsMsg'
Learn More

Hexadiline, TRC

Product Code. 30397606
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397606 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397606 Supplier Toronto Research Chemicals Supplier No. TRCH29424450MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexadiline
CAS 3626-67-3
Molecular Formula C19 H33 N
Quantity 50 mg
Synonym Hexadiline, 2-(2,2-Dicyclohexylethenyl)piperidine, Hexadiline, MRL 38, Hexadylamine
InChI Formula InChI=1S/C19H33N/c1-3-9-16(10-4-1)19(17-11-5-2-6-12-17)15-18-13-7-8-14-20-18/h15-18,20H,1-14H2
SMILES C1CCC(CC1)C(=CC2CCCCN2)C3CCCCC3
IUPAC Name 2-(2,2-dicyclohexylethenyl)piperidine
Molecular Weight (g/mol) 275.472
Formula Weight 275.261
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.