missing translation for 'onlineSavingsMsg'
Learn More

Hexadecyltrimethylammonium Chloride, 97%

Product Code. 10762402 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10762402 5 g Glass Bottle 5g
10465081 25 g Plastic bottle 25g
10636994 100 g Plastic bottle 100g
15199273 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10762402 Supplier Thermo Scientific Acros Supplier No. 411410050
  • ONLINE EXCLUSIVE PRICE
    £58.80
    £52.65 / 5g
    Save £6.15
    10% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 112-02-7 | C19H42ClN | 320.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-02-7
Molecular Formula C19H42ClN
Molecular Weight (g/mol) 320.00
MDL Number MFCD00011773
InChI Key WOWHHFRSBJGXCM-UHFFFAOYSA-M
Synonym hexadecyltrimethylammonium chloride, cetrimonium chloride, cetyltrimethylammonium chloride, n,n,n-trimethylhexadecan-1-aminium chloride, dehyquart a, cetyl trimethyl ammonium chloride, aliquat 6, trimethylcetylammonium chloride, arquad 16-50, n-hexadecyltrimethylammonium chloride
PubChem CID 8154
ChEBI CHEBI:53581
IUPAC Name hexadecyl(trimethyl)azanium;chloride
SMILES [Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Melting Point 234°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass Bottle
Linear Formula CH3(CH2)15N(Cl)(CH3)3
Quantity 5 g
Solubility Information Solubility in water: soluble
Formula Weight 319.75
Percent Purity 97%
Physical Form Amorphous Powder
Chemical Name or Material Hexadecyltrimethylammonium chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexadecyltrimethylammonium chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.