missing translation for 'onlineSavingsMsg'
Learn More

Hexadecyltrimethylammonium Bromide-d3, TRC

Product Code. 30402373
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30402373 1 g -
30391480 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30402373 Supplier Toronto Research Chemicals Supplier No. TRCH2938621G
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1092921-73-7
Molecular Formula C19 2H3 H39 N . Br
Molecular Weight (g/mol) 367.47
Synonym 1-Hexadecanaminium, N,N-dimethyl-N-(methyl-d3)-, bromide (1:1), Hexadecyltrimethylammonium Bromide-d3, Cetrimonium-D3 Bromide (N-methyl-D3)
IUPAC Name hexadecyl-dimethyl-(trideuteriomethyl)azanium;bromide
SMILES [Br-].[2H]C([2H])([2H])[N+](C)(C)CCCCCCCCCCCCCCCC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C19H42N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;/h5-19H2,1-4H3;1H/q+1;/p-1/i2D3;
Formula Weight 366.2689
Chemical Name or Material Hexadecyltrimethylammonium Bromide-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.