missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecyl Acetate, TRC
Specifications
Specifications
| Chemical Name or Material | Hexadecanol acetate |
| CAS | 629-70-9 |
| Molecular Formula | C18 H36 O2 |
| Quantity | 500 mg |
| Synonym | 1-Hexadecanol, acetate (6CI,7CI,8CI,9CI), 1-Acetoxyhexadecane, Cetyl acetate, ENT 1025, Hexadecanol acetate, Hexadecyl acetate, NSC 8492, Palmityl acetate |
| InChI Formula | InChI=1S/C18H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20-18(2)19/h3-17H2,1-2H3 |
| SMILES | CCCCCCCCCCCCCCCCOC(=O)C |
| IUPAC Name | hexadecyl acetate |
| Molecular Weight (g/mol) | 284.48 |
| Formula Weight | 284.2715 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?