missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecanoic-11,11-d2 Acid, CDN
Specifications
Specifications
| Chemical Name or Material | Hexadecanoic-11,11-D2 Acid |
| CAS | 1219805-64-7 |
| Molecular Formula | C16 D2 H30 O2 |
| Quantity | 0.25 g |
| Synonym | Hexadecanoic-11,11-d2 acid, Palmitic-11,11-d2 acid, 11,11-Dideuteriohexadecanoic acid, 11,11-D2-Palmitic acid, Hexadecanoic-11,11-D2 Acid, [11,11-2H2]Hexadecanoic acid, [11,11-2H2]Palmitic acid, Hexadecanoic Acid-11,11-d2 |
| InChI Formula | InChI=1 S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i6D2 |
| SMILES | [2 H]C([2 H])(CCCCC)CCCCCCCCCC(=O)O |
| IUPAC Name | 11,11-dideuteriohexadecanoic acid |
| Molecular Weight (g/mol) | 258.4364 |
| Formula Weight | 258.2528 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?