missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Hexadecanedioic acid
Specifications
Specifications
| Content And Storage | Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month |
| Product Type | Natural Products |
| Quantity | 100 mg |
| Shipping Condition | Room temperature |
| CAS | 505-54-4 |
| Molecular Weight (g/mol) | 286.41 g/mol |
| Solubility | DMSO : 100 mg/mL (349.15 mM; Need ultrasonic) |
| Target | NO Synthase ∣ MMP ∣ Endogenous Metabolite |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?