missing translation for 'onlineSavingsMsg'
Learn More

Hexadecane, TRC

Product Code. 30137791
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137791 500 mL 500mL
17933475 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 30137791 Supplier Toronto Research Chemicals Supplier No. TRCH293630500ML
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-76-3
Molecular Formula C16 H34
Molecular Weight (g/mol) 226.44
Synonym Hexadecane, Cetane, NSC 7334, S 6 (alkane), n-Cetane, n-Hexadecane
IUPAC Name hexadecane
SMILES CCCCCCCCCCCCCCCC

Specifications

Quantity 500 mL
InChI Formula InChI=1S/C16H34/c1-3-5-7-9-11-13-15-16-14-12-10-8-6-4-2/h3-16H2,1-2H3
Formula Weight 226.2661
Chemical Name or Material n-Hexadecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.