missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexacosanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Hexacosanoic acid (Cerotic acid) |
| CAS | 506-46-7 |
| Molecular Formula | C26 H52 O2 |
| Quantity | 250 mg |
| Synonym | Hexacosanoic acid (8CI, 9CI, ACI), Cerotic acid (6CI, 7CI), Ceratinic acid, Ceric acid, Cerinic acid, Cerylic acid, n-Hexacosanoic acid, NSC 4205 |
| InChI Formula | InChI=1S/C26H52O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26(27)28/h2-25H2,1H3,(H,27,28) |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O |
| IUPAC Name | hexacosanoic acid |
| Molecular Weight (g/mol) | 396.69 |
| Formula Weight | 396.3967 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?