missing translation for 'onlineSavingsMsg'
Learn More

Hexacosanoic Acid, TRC

Product Code. 30391461
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30391461 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30391461

Brand: Toronto Research Chemicals TRCH2909101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexacosanoic acid (Cerotic acid)
CAS 506-46-7
Molecular Formula C26 H52 O2
Quantity 1 g
Synonym Hexacosanoic acid (8CI, 9CI, ACI), Cerotic acid (6CI, 7CI), Ceratinic acid, Ceric acid, Cerinic acid, Cerylic acid, n-Hexacosanoic acid, NSC 4205
InChI Formula InChI=1S/C26H52O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26(27)28/h2-25H2,1H3,(H,27,28)
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O
IUPAC Name hexacosanoic acid
Molecular Weight (g/mol) 396.69
Formula Weight 396.3967
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.