missing translation for 'onlineSavingsMsg'
Learn More

Hexacosanoic acid, MedChemExpress

Product Code. 16411047
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity unitSize
16411047 50 mg 50mg
1 options
This item is not returnable. View return policy

Product Code. 16411047

Brand: Medchem Express HY113301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hexacosanoic acid is a long-chain fatty acid related to various diseases such as adrenoleukodystrophy (ALD), adrenomyeloneuropathy (AMN) and atherosclerosis.

Hexacosanoic acid is a long-chain fatty acid related to various diseases such as adrenoleukodystrophy (ALD), adrenomyeloneuropathy (AMN) and atherosclerosis.

Specifications

Chemical Name or Material Hexacosanoic acid
CAS 506-46-7
Color White
Purity Grade Notes Research
Molecular Formula C26H52O2
Quantity 50 mg
Solubility Information THF : 5.56 mg/mL (14.02 mM; Need ultrasonic)
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 396.69
Formula Weight 396.69
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.