missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexacosanoic-12,12,13,13-d4 Acid, CDN
Specifications
Specifications
| Chemical Name or Material | Hexacosanoic Acid-d4 |
| CAS | 1194984-85-4 |
| Molecular Formula | C26 D4 H48 O2 |
| Quantity | 0.05 g |
| Synonym | Cerotic Acid-d4, Ceratinic Acid-d4, Ceric Acid-d4, Cerinic Acid-d4, Cerylic Acid-d4, NSC 4205-d4, n-Hexacosanoic Acid-d4, Hexacosanoic-12,12,13,13-d4 Acid |
| InChI Formula | InChI=1 S/C26H52O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26(27)28/h2-25H2,1H3,(H,27,28)/i14D2,15D2 |
| SMILES | [2 H]C([2 H])(CCCCCCCCCCCCC)C([2 H])([2 H])CCCCCCCCCCC(=O)O |
| IUPAC Name | 12,12,13,13-tetradeuteriohexacosanoic acid |
| Molecular Weight (g/mol) | 400.71 |
| Formula Weight | 400.4218 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?