missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexacosane, 99%, Thermo Scientific Chemicals
£30.70 - £88.60
Chemical Identifiers
CAS | 630-01-3 |
---|---|
Molecular Formula | C26H54 |
Molecular Weight (g/mol) | 366.72 |
MDL Number | MFCD00009354 |
InChI Key | HMSWAIKSFDFLKN-UHFFFAOYSA-N |
Synonym | n-hexacosane, pentacosane, methyl, ch3-ch2 24-ch3, hexacosane, analytical standard, hexacosane, acmc-1b86y |
PubChem CID | 12407 |
ChEBI | CHEBI:32940 |
IUPAC Name | hexacosane |
SMILES | CCCCCCCCCCCCCCCCCCCCCCCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10082940
|
Thermo Scientific Acros
221220010 |
1 g | Glass bottle |
£30.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10667362
|
Thermo Scientific Acros
221220050 |
5 g | Glass bottle |
£88.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
630-01-3 | |
366.72 | |
HMSWAIKSFDFLKN-UHFFFAOYSA-N | |
12407 | |
hexacosane |
C26H54 | |
MFCD00009354 | |
n-hexacosane, pentacosane, methyl, ch3-ch2 24-ch3, hexacosane, analytical standard, hexacosane, acmc-1b86y | |
CHEBI:32940 | |
CCCCCCCCCCCCCCCCCCCCCCCCCC |
Specifications
630-01-3 | |
White | |
Authentic | |
C26H54 | |
MFCD00009354 | |
n-hexacosane, pentacosane, methyl, ch3-ch2 24-ch3, hexacosane, analytical standard, hexacosane, acmc-1b86y | |
CCCCCCCCCCCCCCCCCCCCCCCCCC | |
366.72 | |
CHEBI:32940 | |
≥98.5% | |
Hexacosane, 99% |
56°C to 59°C | |
0.8000g/mL | |
99% | |
CH3(CH2)24CH3 | |
01, 175 | |
HMSWAIKSFDFLKN-UHFFFAOYSA-N | |
hexacosane | |
12407 | |
366.7 | |
Crystalline Solid |
Safety and Handling
missing translation for 'einecsNumber' : 211-124-1
missing translation for 'tsca' : TSCA
RUO – Research Use Only