missing translation for 'onlineSavingsMsg'
Learn More

Hexachloroethane, SPEX CertiPrep™

Product Code. 12949264 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
12949264 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 12949264

Brand: SPEX Certiprep S2155

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Hexachloroethane67-72-11000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging 2mL amber prescored glass ampule
CAS 67-72-1, 67-56-1
CAS Min % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molecular Formula C2Cl6
MDL Number MFCD00000799
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key VHHHONWQHHHLTI-UHFFFAOYSA-N
Type Certified Reference Material
SMILES ClC(Cl)(Cl)C(Cl)(Cl)Cl
IUPAC Name hexachloroethane
Solvent or Matrix Methanol-P and T
Molecular Weight (g/mol) 236.72
PubChem CID 6214
Quantity 1 mL
ChEBI CHEBI:39227
Chemical Name or Material Hexachloroethane
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.