missing translation for 'onlineSavingsMsg'
Learn More

Hexachloroethane, 99%

Product Code. 11349095
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11349095 100 g 100g
11353446 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349095 Supplier Thermo Scientific Alfa Aesar Supplier No. L14297.22
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 67-72-1 | C2Cl6 | 236.72 g/mol

Hexachloroethane is used as catalyst in preparation of chlorosilanes from hydrosilanes. It is also used to investigate the transformation of hexachloroethane in natural waters and as reagent during polycondensation reaction of p-aminobenzoic acid. It is used as a source of chlorine in the chlorodehydroxylation of alcohols in the presence of triphenylphosphine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexachloroethane is used as catalyst in preparation of chlorosilanes from hydrosilanes. It is also used to investigate the transformation of hexachloroethane in natural waters and as reagent during polycondensation reaction of p-aminobenzoic acid. It is used as a source of chlorine in the chlorodehydroxylation of alcohols in the presence of triphenylphosphine.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-72-1
Molecular Formula C2Cl6
Molecular Weight (g/mol) 236.72
MDL Number MFCD00000799
InChI Key VHHHONWQHHHLTI-UHFFFAOYSA-N
Synonym hexachloroethane, perchloroethane, ethane, hexachloro, avlothane, carbon hexachloride, hexachlorethane, mottenhexe, distokal, distopan, distopin
PubChem CID 6214
ChEBI CHEBI:39227
SMILES ClC(Cl)(Cl)C(Cl)(Cl)Cl

Specifications

Melting Point 185°C to 190°C (sublimation)
Density 2.091
Odor Camphor-like
Quantity 100 g
UN Number UN3077
Beilstein 1740341
Merck Index 14,4679
Solubility Information Insoluble in water.
IUPAC Name hexachloroethane
Formula Weight 236.74
Percent Purity 99%
Chemical Name or Material Hexachloroethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.