missing translation for 'onlineSavingsMsg'
Learn More

Hexachlorobenzene, TRC

Product Code. 30137449
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137449 100 g 100g
18020840 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137449 Supplier Toronto Research Chemicals Supplier No. TRCH290815100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-74-1
Molecular Formula C6 Cl6
Molecular Weight (g/mol) 284.78
Synonym Benzene, 1,2,3,4,5,6-hexachloro-, Benzene, hexachloro- (8CI,9CI), 1,2,3,4,5,6-Hexachlorobenzene, Amatin, Anticarie, BHC, Bunt-cure, Bunt-no-more, Co-op Hexa, HCB, HCB (pesticide), Hexachlorobenzene, Julin's carbon chloride, NSC 9243, No Bunt, No Bunt 40, No Bunt 80, No Bunt Liquid, Pentachlorophenyl chloride, Perchlorobenzene, Sanocide, Snieciotox, Zaprawa nasienna sneciotox
IUPAC Name 1,2,3,4,5,6-hexachlorobenzene
SMILES Clc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl

Specifications

Quantity 100 g
InChI Formula InChI=1S/C6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9
Formula Weight 281.8131
Chemical Name or Material Hexachlorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.