missing translation for 'onlineSavingsMsg'
Learn More

Hexachloroacetone, 99%

Product Code. 11478890
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478890 100 g 100g
11490857 250 g 250g
11480857 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478890 Supplier Thermo Scientific Alfa Aesar Supplier No. B23259.22
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 116-16-5 | C3Cl6O | 264.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-16-5
Molecular Formula C3Cl6O
Molecular Weight (g/mol) 264.73
MDL Number MFCD00000796
InChI Key DOJXGHGHTWFZHK-UHFFFAOYSA-N
Synonym hexachloroacetone, hexachloro-2-propanone, perchloroacetone, hexachloropropanone, kureha hca, hca weedkiller, perchloro-2-propanone, bis trichloromethyl ketone, acetone, hexachloro, 2-propanone, hexachloro
PubChem CID 8303
ChEBI CHEBI:82243
IUPAC Name 1,1,1,3,3,3-hexachloropropan-2-one
SMILES ClC(Cl)(Cl)C(=O)C(Cl)(Cl)Cl

Specifications

Melting Point -2°C
Boiling Point 203°C to 205°C
Flash Point >110°C (230°F)
Quantity 100 g
UN Number UN2661
Beilstein 1707475
Formula Weight 264.75
Percent Purity 99%
Chemical Name or Material Hexachloroacetone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.