missing translation for 'onlineSavingsMsg'
Learn More

Hexacarbonyltungsten, 97%

Product Code. 11379268
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379268 50 g 50g
11389268 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379268 Supplier Thermo Scientific Alfa Aesar Supplier No. 043205.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 14040-11-0 | C6O6W

Hexacarbonyltungsten is used as a precursor to a Fischer carbine complex, which used in the hiyama cross-coupling reactions. It is widely used in electron beam-induced deposition technique as well as a source of tungsten atoms. It is utilized to desulfurize organosulfur compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14040-11-0
Molecular Formula C6O6W
MDL Number MFCD00011462
Synonym Tungsten hexacarbonyl

Specifications

Melting Point 172°C to 174°C
Color White
Density 2.65
Assay Percent Range 97%
Odor Nearly odorless
Quantity 50 g
Solubility Information Slightly soluble in tetrahydrofuran. Insoluble in water.
Formula Weight 351.91
Vapor Pressure 1.596hPa (1mm Hg) at 67°C
Physical Form Crystalline
Chemical Name or Material Hexacarbonyltungsten
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.