missing translation for 'onlineSavingsMsg'
Learn More

Hexacarbonylmolybdenum, 98%

Product Code. 11362138
Change view
Click to view available options
Quantity:
2.5 g
25 g
100 g
Unit Size:
100g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362138 100 g 100g
15421037 2.5 g 2.5g
11352138 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11362138 Supplier Thermo Scientific Alfa Aesar Supplier No. 013057.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13939-06-5 | C6MoO6 | 264.01 g/mol

Hexacarbonylmolybdenum is widely utilized in electron beam-induced deposition technique providing an appropriate source of molybdenum atoms. It finds an application as important reagent in organometallic synthesis because of replacement of carbonyl ligands by other donor ligands. Its derivative is act as a catalyst in organic synthesis, which is used in alkyne metathesis and the Pauson-Khand reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13939-06-5
Molecular Formula C6MoO6
Molecular Weight (g/mol) 264.01
MDL Number MFCD00003466
InChI Key KMKBZNSIJQWHJA-UHFFFAOYSA-N
Synonym molybdenumhexacarbonyl, carbon monooxide-molybdenum 6:1, hexakis carbon monoxide molybdenum, hexacarbonylmolybdenu, hexakis methanidylidyneoxidanium molybdenum, hexacarbonylmolybdenum, may contain up to cr 5g
PubChem CID 2724068
ChEBI CHEBI:30508
IUPAC Name carbon monoxide;molybdenum
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Mo]

Specifications

Melting Point ∽150°C (decomposition)
Color White
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 98%
Linear Formula Mo(CO)6
UN Number UN3466
Solubility Information Slightly soluble in tetrahydrofuran,diglyme and acetonitrile.
Formula Weight 264
Percent Purity 98%
Concentration or Composition (by Analyte or Components) May contain up to 1% Cr
Density 1.96g/mL
Chemical Name or Material Hexacarbonylmolybdenum, Up to 1% Cr
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.