missing translation for 'onlineSavingsMsg'
Learn More

Hexabromocyclododecane (Mixture of Diastereomers), TRC

Product Code. 30399910
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399910 5 g
30407717 10 g
30397743 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399910 Supplier Toronto Research Chemicals Supplier No. TRCH2907205G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25637-99-4
Molecular Formula C12H18Br6
Molecular Weight (g/mol) 641.7
Synonym gamma-Hexabromocyclododecane, β-Hexabromocyclododecane, α-Hexabromocyclododecane, 1,2,5,6,9,10-Hexabromocyclododecane, Hexabromocyclododecane
IUPAC Name 1,3,5,7,9,11-hexabromocyclododecane
SMILES BrC1CC(Br)CC(Br)CC(Br)CC(Br)CC(Br)C1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H18Br6/c13-7-1-8(14)3-10(16)5-12(18)6-11(17)4-9(15)2-7/h7-12H,1-6H2
Formula Weight 635.65
Chemical Name or Material Hexabromocyclododecane (mix of isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.