missing translation for 'onlineSavingsMsg'
Learn More

Hexaammineruthenium(III) chloride, 98%

Product Code. p-3735332 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10583884

Brand: Thermo Scientific Acros 363342500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14282-91-8 | Cl3H18N6Ru | 309.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14282-91-8
Molecular Formula Cl3H18N6Ru
Molecular Weight (g/mol) 309.61
MDL Number MFCD00011478
InChI Key GBDZMMXUOBAJMN-UHFFFAOYSA-K
Synonym hexaammineruthenium iii chloride, hexaammineruthenium trichloride, hexaamminetrichlororuthenium, hexaammineruthenium trichloride, hydrate, ruthenium 3+ , hexaammine-, trichloride, hydrate, ruthenium 3+ chloride ammoniate 1:3:6, hexaammineruthenium iii trichloride, ruthenium 3+ ion hexaamine trichloride, ruthenium 3+ , hexaammine-, trichloride
PubChem CID 159731
SMILES N.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Ru+3]

Specifications

Color Yellow
Physical Form Crystalline Powder
Quantity 250 mg
Assay Percent Range 98%
Solubility Information Solubility in water: soluble
IUPAC Name azane;ruthenium(3+);trichloride
Formula Weight 309.61
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Hexaammineruthenium(III) chloride
compliance-icons
Product Identifier
  • Hexaammineruthenium(III) chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.