missing translation for 'onlineSavingsMsg'
Learn More

Hexaamminenickel(II) bromide, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11400830
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400830 25 g 25g
11410830 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11400830 Supplier Thermo Scientific Alfa Aesar Supplier No. 044482.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13601-55-3
Molecular Formula Ni(NH3)6Br2
MDL Number MFCD04039935
Synonym hexaamminenickel ii bromide, dibromonickel hexaamine

Specifications

Quantity 25 g
Formula Weight 320.68
Percent Purity 99.999%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Hexaamminenickel (II) bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.