missing translation for 'onlineSavingsMsg'
Learn More

Hexaamminecobalt(III) chloride, 97%, Thermo Scientific Chemicals

Product Code. 11371309
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Hexaamminecobalt(III) chloride is used in X-ray crystallography and NMR. It is useful in DNA condensation studies. It is suitable for induction of transitions of DNA from B to A or Z forms, induction of nucleic acid crystal growth, stabilization of tertiary tRNA interactions, preparation of ligation buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10534-89-1
Molecular Formula [Co(NH3)6]Cl3
MDL Number MFCD00036304

Specifications

Color Orange
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 97%
Solubility Information Soluble in ammonia.
Formula Weight 267.48
Odor Odorless
Appearance Orange powder
Density 1.71 g/cm3
Chemical Name or Material Hexaamminecobalt(III) chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'